Natriumperkloraattimonohydraatii ≥99.0%, HiPerSolv CHROMANORM® for HPLC

Toimittaja: VWR Chemicals
Vaara

Synonyms: Perchloric acid sodium salt

153233MEA 181 EUR
153233M
Natriumperkloraattimonohydraatii ≥99.0%, HiPerSolv CHROMANORM® for HPLC
Natriumperkloraattimonohydraatii
Kaava: NaClO₄·H₂O
MW: 140.46 g/mol
Sulamispiste: 130
Tiheys: 2.02 g/cm³ (25 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00149164
CAS-numero: 7791-07-3
EINECS: 231-511-9
UN: 1502
ADR: 5.1 II
Merck Index: 12,08798

Order Now

Testitulosten erittely

Assay (argentometric, as monohydrate) Min. 99.0 %
Transmittance (254 nm) (1 mol/l) Min. 96 %
Chloride, chlorate (as Cl) Max. 0.002 %
SO4 (Sulphate) Max. 0.002 %
Ca (Calcium) Max. 0.002 %
Fe (Iron) Max. 0.0003 %
K (Potassium) Max. 0.005 %
Pb (Lead) Max. 0.0005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR