Europiumtrinitraatti-6 H2O ≥99.9% (REO, rare earth oxide basis), REacton®

Toimittaja: Thermo Fisher Scientific
Vaara

15290.18
15290.18EA 2810 EUR
15290.18
Europiumtrinitraatti-6 H2O ≥99.9% (REO, rare earth oxide basis), REacton®
Europiumtrinitraatti-6 H2O

Purity: 99.9% CAS No.: 10031-53-5 Molecular Formula: Eu(NO3)3.6H2O Formula Weight: 446.06 (337.97anhy) Physical Form: Crystalline or powder Appearance: White Odour: Odourless Melting Point: 85° Merck Reference: 14,3904 UN No.: 1477 MDL No.: MFCD00149700

Kaava: EuN₃O₉·6H₂O
Sulamispiste: 85 °C
Tiheys: 2,258 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00149700
CAS-numero: 10031-53-5
EINECS: 233-380-3
UN: 1477
ADR: 5.1,III
Merck Index: 14,3904

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR