Glysiini, BAKER ANALYZED® for biotechnology, J.T.Baker®

Toimittaja: Avantor

1504.0100EA 25.27 EUR
1504.0100 1504.0500
Glysiini, BAKER ANALYZED® for biotechnology, J.T.Baker®
Glysiini
Kaava: NH₂CH₂COOH
Kiehumispiste: 181 °C (1013 hPa)
Sulamispiste: 83 °C
Tiheys: 1,64 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00008131
CAS-numero: 56-40-6
EINECS: 200-272-2
Merck Index: 13,04504
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d944007d30d5d4650062014a1b152bd9bfb66ea3&vid=093f4227cbdd1c4c6515ef9c4b85d93799877dcc&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Assay (by Perchloric Acid titrn.) min. 99 %
Heavy Metals (as Pb) max. 0.002 %
Homogenity (by TLC) passes test
Loss on Drying at 105°C max. 0.2 %
Readily Carbonisable Substances passes test
Residue after Ignition max. 0.1 %
Trace Impurities (in ppm) Arsenic (As) max. 3
Trace Impurities (in ppm) Iron (Fe) max. 5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR