Regenerating salts for dishwashers
Toimittaja: KAMMERER
1161340
148-2501EA 8.3 EUR
148-2501
Regenerating salts for dishwashers
Puhdistusaineet Pesuaineet ja pintojen puhdistusaineet
Regenerating salts for water softening systems for dishwashers, it protects dishes and machines from calcification. Also they ensures optimum efficiency of the dish cleaner and are meant for all cleaning systems.
  • Highly pure and residue-free soluble
  • Coarse-grained for optimal solubility
Sertifikaatit: EN 973 Type A.