Heavy Metals, plasma standard solution, Ag, As, Ba, Cd, Cr, Hg, Pb, Se @ 100µg/ml, Specpure®

Toimittaja: Thermo Fisher Scientific

14657.AE 14657.AP
14657.AEEA 449 EUR
14657.AE 14657.AP
Heavy Metals, plasma standard solution, Ag, As, Ba, Cd, Cr, Hg, Pb, Se @ 100µg/ml, Specpure®
Standardiaineet ja vertailumateriaalit Cationic Standards Multi-Element Standards

UN No.: UN3264
MDL No.: MFCD02091713

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR