Kupari(I)kloridi ≥99.999% (metals basis)

Toimittaja: Thermo Fisher Scientific
Varoitus

Synonyms: Kuparikloridi

14644.06 14644.14 14644.22
14644.06EA 144 EUR
14644.06 14644.14 14644.22
Kupari(I)kloridi ≥99.999% (metals basis)
Kupari(I)kloridi

Purity: 99.999% CAS No.: 7758-89-6 Molecular Formula: CuCl Formula Weight: 98.99 Physical Form: Powder Density: 4.14 Refractive Index: 1.93 Sensitivity: Air and Moisture Sensitive Appearance: White Melting Point: 430° Boiling Point: 1490° Merck Reference: 14,2660 UN No.: 2802 MDL No.: MFCD00010971

Kaava: ClCu
MW: 99 g/mol
Kiehumispiste: 1490 °C
Sulamispiste: 430
Tiheys: 4.14
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00010971
CAS-numero: 7758-89-6
EINECS: 231-842-9
UN: 2802
ADR: 8 III
Merck Index: 14,2660

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR