Dysprosium(III) acetate tetrahydrate ≥99.9% (REO, rare earth oxide basis), REacton®

Toimittaja: Thermo Fisher Scientific

14595.14 14595.22
14595.14EA 180 EUR
14595.14 14595.22
Dysprosium(III) acetate tetrahydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Dysprosium(III) acetate tetrahydrate

Purity: 99.9% CAS No.: 15280-55-4 Molecular Formula: Dy(OOCCH)3.4H2O Formula Weight: 411.68 (339.62anhy) Physical Form: Crystalline Aggregates Odour: Odourless MDL No.: MFCD00013033

Kaava: C₆H₉DyO₆·4H₂O
Sulamispiste: 120 °C (haj. lämpötila)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00013033
CAS-numero: 15280-55-4
EINECS: 242-565-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR