Kupari(II)sulfiitti, ultra dry ≥99.98% (metals basis)

Toimittaja: Thermo Fisher Scientific


Ampuled under argon

14300.09 14300.18
14300.09EA 577 EUR
14300.09 14300.18
Kupari(II)sulfiitti, ultra dry ≥99.98% (metals basis)
Kupari(II)sulfiitti

Purity: 99.98% CAS No.: 1317-40-4 Molecular Formula: CuS Formula Weight: 95.60 Physical Form: Powder, Ampouled under argon Density: 3.9-4.6 Refractive Index: 1.45 Sensitivity: Air and Moisture Sensitive Odour: Odourless Melting Point: 220° dec. Merck Reference: 14,2965 MDL No.: MFCD00016066

Kaava: CuS
Tiheys: 4,6
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00016066
CAS-numero: 1317-40-4
EINECS: 215-271-2
Merck Index: 14,2965

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR