Titaani standard dissolved in oil, 1.000 µg/g Ti in mineral oil Specpure® for wear metal analysis

Toimittaja: Thermo Fisher Scientific

14204.1Y 14204.18
14204.1YEA 0 EUR
14204.1Y 14204.18
Titaani standard dissolved in oil, 1.000 µg/g Ti in mineral oil Specpure® for wear metal analysis
Standardiaineet ja vertailumateriaalit Cationic Standards Titanium Standards

Physical Form: Liquid
MDL No.: MFCD00011264

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR