Ultrasonic baths, SONOREX SUPER
Toimittaja: BANDELIN-ELECTRONIC
311 164 329 301 312 303 326 305 646 316 321 263 322 324 323 327 795 277 207 306 319 661 3066 306-GB 329-CH 329-GB
142-1100EA 912 EUR
142-1100 142-1110 142-1112 142-1120 142-1130 142-1210 142-1220 142-1235 142-1236 142-1240 142-1250 142-1260 142-1265 142-1266 142-1280 142-1399 142-1400 142-1401 142-1402 510-7120 BAND319 BAND661 BAND3066 BAND306-GB BAND329-CH BAND329-GB
Ultrasonic baths, SONOREX SUPER
Ultraäänihauteet
SONOREX SUPER is the high-power ultrasonic bath for aqueous cleaning solutions.
  • H-types with built-in heating from 20 to 80 °C, thermostatically adjustable
  • Oscillating tank of stainless steel, RK 102 H stainless steel, hard chromium plated
  • Timer 1 - 15 minutes and continuous operation
  • Housing of stainless steel - drip proof
  • Drain valve for simple emptying, one-piece welded drain bow