Ultrasonic baths, digital, XUB Series
Toimittaja: GRANT INSTRUMENTS
XUB5 XUB25 XUB12 XUB18
142-0149EA 2540 EUR
142-0149 142-0148 142-0124 142-0147
Ultrasonic baths, digital, XUB Series
Ultraäänihauteet
XUB range of digitally controlled benchtop ultrasonic baths offer consistent and reliable performance in a variety of environments such as laboratory, scientific and healthcare. Incorporating frequency LEAP technology to ensure uniform levels of ultrasonic activity throughout the fluid, these baths offer vastly improved performance giving a more accurate and precise ultrasonic process.
  • Frequency LEAP technology provides more homogeneous ultrasonic activity throughout the tank, reducing dead spots and standing waves
  • Stainless steel basket - designed