Ultrasonic baths, digital, Bransonic®
Toimittaja: BRANSON
CPX-952-838R CPX-952-539R CPX-952-538R CPX-952-339R CPX-952-239R CPX-952-238R CPX-952-139R CPX-952-138R CPX-952-338R CPX-952-839R
142-0141EA 4140 EUR
142-0141 142-0138 142-0117 142-0135 142-0132 142-0115 142-0129 142-0114 142-0116 142-0142
Ultrasonic baths, digital, Bransonic®
Ultraäänihauteet
CPX ultrasonic baths feature digital controls with power tracking capabilities to adjust for light or heavy loads, high/low power control, degassing and sleep mode.
  • Plastic body, chemical-resistant
  • Sweep frequency eliminates standing waves and creates consistent cavitation throughout the tank
  • 40 kHz rugged industrial transducers
  • 99-min digital timer with continuous ultrasonic operation
  • Controls are located above and behind the cleaning tank in an easily accessible control tower panel, and raised above the tank to avoid damage from cleaning solutions
  • When