Dinatriummetasilikaatti, vedetön, granular, tech. –18 Mesh

Toimittaja: Thermo Fisher Scientific
Vaara

14106.A1 14106.D9 14106.99
14106.A1EA 149 EUR
14106.A1 14106.D9 14106.99
Dinatriummetasilikaatti, vedetön, granular, tech. –18 Mesh
Dinatriummetasilikaatti

Sodium metasilicate is used as a starting material for zeolites and silica catalysts. It acts as an adhesive and binder, corrosion inhibitor, penetrating sealant, coagulant in waste water treatment, fixative for hand dyeing with reactive dye and detergent auxiliaries.


It is also used in boiler compounds, cleaners, bleaches, aluminum paint strippers and washing carbonated drinking bottles. It finds application in cements, drilling fluids, fireproofing wood, refractories and automobiles. Further, it serves as a surfactant, emulsifier, wetting agent, suspending agent, dispersing agent and anti-corrosive agent in boiler water.


Incompatible with acids, bronze, aluminum, brass, copper, lead, tin, zinc and other alkali sensitive metals.

Kaava: Na₂O₃Si
Sulamispiste: 1088 °C
Tiheys: 2,61
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00003492
CAS-numero: 6834-92-0
EINECS: 229-912-9
UN: 3253
ADR: 8,III
Merck Index: 14,8642

Order Now

Testitulosten erittely

Assay (Unspecified), SiO₂ 44.7 - 47.6%
Assay (Unspecified), Na₂O 49.1 - 51.7%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR