SlimLine PLW 7111 Laboratory Glasswashers
Toimittaja: MIELE
11875090 11875120 11875190 11875200 11875410 11875470
141-1596EA 28963 EUR
141-1596 141-1597 141-1598 141-1599 141-1600 141-1601
SlimLine PLW 7111 Laboratory Glasswashers
Laboratoriolasinpesukoneet
The PLW 7111 SlimLine laboratory washers offer a comprehensive and systematic solution for analytically pure reprocessing of laboratory glassware.
  • Smart Load Plus - for flexible use of wash cabinet space
  • EasyLoad - systematic efficiency and convenience
  • Space-saving SlimLine design
  • Freely programmable controls
  • Flexible reprocessing of laboratory glassware
  • Status display via chamber lighting
  • Connectivity-ready
The PLW 7111 series of SlimLine laboratory glasswashers provide performance and user-friendliness. This entirely redesigned washing system boastsMore Product Information