VWR® Bucket Liners, Sterile
Toimittaja: VWR Collection
141-1380EA 205 EUR
141-1380 141-1443
VWR® Bucket Liners, Sterile
Pussit ja sulkijat Jätepussit, -säkit
Bucket liners create an impermeable barrier between the disinfectant and receptacle.
  • LDPE/nylon blend
  • Irradiated
  • Fits B-7 series buckets
Helps ensure the integrity of a disinfectant blend.
Pakkaus: Individually bagged, double-bagged in groups of 6. 4 packs of 6 per case.