QuickTask™ flat mops
Toimittaja: CONTEC
2645 2642 2643 2725E 2740 TASK0400 2686 TASK0300 TASK0200 TASK0250 2746 TASK0500 TASK0550 2699P 2906
129-0205EA 523 EUR
129-0205 129-0202 129-0203 141-0960 141-0961 141-0978 129-0573 141-0977 141-0975 141-0976 141-1367 141-0979 141-0980 CONT2699P CONT2906
QuickTask™ flat mops
Siivousvälineet
QuickTask™ mopping system. The QuickTask™ product line consists of a variety of knitted polyester, polyester/cellulose, microfibre/foam and microfibre flat mop heads that are easily installed onto a hinged mop head frame and QuickConnect interchangeable handle. QuickTask™ mop head frames are available in stainless steel or durable, autoclavable polypropylene.
  • All mop head covers are compatible with a wide range of chemicals and disinfectants
  • Ideally suited for cleaning floors, including perforated cleanroom floors and grates
  • TASK0300 is ideal for effective