Mops and handles
Toimittaja: Rubbermaid Commercial Products
R034562 FGQ75000YL00 FGQ760000000
141-0121EA 43.9 EUR
141-0121 141-0126 141-0127
Mops and handles
Siivousvälineet
Microfibre mops:
High absorbency microfibre mop: Microfibre delivers superior cleaning performance and durability. Looped fringe mop absorbs up to 0,6 litres of liquid.
Microfibre dry mop: 100% microfibre collects and holds dirt electrostatically for superior dusting results.
For dry dusting only — do not use wet.
Quick-Connect handle HYGEN™:
Available as straight or ergonomically bent styles, in fixed and adjustable lengths. Rubbermaid HYGEN™ handles contain non magnetic aluminium and are safe for use in MRI rooms.
Lightweight aluminium construction for improved productivity
User friendlyMore Product Information