Bariumfluoridi, Optical Grade

Toimittaja: Thermo Fisher Scientific
Varoitus

14057.06 14057.14 14057.22
14057.06EA 102 EUR
14057.06 14057.14 14057.22
Bariumfluoridi, Optical Grade
Bariumfluoridi

CAS No.: 7787-32-8 Molecular Formula: BaF2 Formula Weight: 175.36 Physical Form: Broken Crystals Density: 4.83 Refractive Index: 1.4741 Odour: Odourless Melting Point: 1353° Boiling Point: 2260° Merck Reference: 14,974 UN No.: 1564 MDL No.: MFCD00003450

Kaava: BaF₂
Kiehumispiste: 2260 °C
Sulamispiste: 1353 °C
Tiheys: 4,83
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00003450
CAS-numero: 7787-32-8
EINECS: 232-108-0
UN: 1564
ADR: 6.1,III
Merck Index: 14,974

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR