Potassium stannate trihydrate ≥95% (metals basis)

Toimittaja: Thermo Fisher Scientific

14015.30 14015.A1
14015.30EA 93.3 EUR
14015.30 14015.A1
Potassium stannate trihydrate ≥95% (metals basis)
Potassium stannate trihydrate

Purity: 95+% CAS No.: 12125-03-0 Molecular Formula: K2SnO3.3H2O Formula Weight: 298.94 (244.89anhy) Physical Form: Crystalline Odour: Odourless MDL No.: MFCD00150391

Kaava: K₂O₃Sn·3H₂O
Sulamispiste: 140 °C (haj. lämpötila)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00150391
CAS-numero: 12125-03-0
EINECS: 235-255-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR