Kuljetusväline, Nalgene®
Toimittaja: Thermo Fisher Scientific
7131-0001
139-3120EA 134 EUR
139-3120
Kuljetusväline, Nalgene®
Kuljetuskorit ja turvakuljetus Kuljetustelineet
HDPE. Foam inserts.
  • For up to six bottles with Ø of 60, 89 and 108 mm
  • Material does not break or splinter
  • Stackable for space-saving
These carriers are suitable for the safe transport of glass and plastic bottles, containers and many other types of laboratory equipment. Two foam inserts with a total of six openings keep bottles apart when the basket is used for bottles.
Colour: translucent.