Vanadiini 99,8 Metals basis, foil, Thickness: 0.127 mm (0.005 in)

Toimittaja: Thermo Fisher Scientific

13783.FY 13783.GK
13783.FYEA 373 EUR
13783.FY 13783.GK
Vanadiini 99,8 Metals basis, foil, Thickness: 0.127 mm (0.005 in)
Vanadiini

Purity: 99.8% CAS No.: 7440-62-2 Physical Form: ≈3.87g/50x100mm Odour: Odourless MDL No.: MFCD00011453

Kaava: V
MW: 50.94 g/mol
Kiehumispiste: 3477
Sulamispiste: 1896
Tiheys: 6.015 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011453
CAS-numero: 7440-62-2
EINECS: 231-171-1
Merck Index: 12,10053

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR