Neodyymitrifluoridi, vedetön ≥99.9% (REO, rare earth oxide basis), REacton®

Toimittaja: Thermo Fisher Scientific
Varoitus

13661.14 13661.22
13661.14EA 304 EUR
13661.14 13661.22
Neodyymitrifluoridi, vedetön ≥99.9% (REO, rare earth oxide basis), REacton®
Neodyymitrifluoridi

Purity: 99.9% CAS No.: 13709-42-7 Molecular Formula: NdF3 Formula Weight: 201.24 Physical Form: Crystalline Lump Density: 6.5 Appearance: Pale purple Odour: Odourless Melting Point: 1410° MDL No.: MFCD00011135

Kaava: F₃Nd
Kiehumispiste: 2327 °C (1013 hPa)
Sulamispiste: 1410 °C
Tiheys: 6,5
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011135
CAS-numero: 13709-42-7
EINECS: 237-253-3
UN: 3288
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR