Tulium(III)fluoridi, vedetön ≥99.9% (REO, rare earth oxide basis)

Toimittaja: Thermo Fisher Scientific
Vaara

13652.06 13652.14
13652.06EA 608 EUR
13652.06 13652.14
Tulium(III)fluoridi, vedetön ≥99.9% (REO, rare earth oxide basis)
Tulium(III)fluoridi

Purity: 99.9% CAS No.: 13760-79-7 Molecular Formula: TmF3 Formula Weight: 225.93 Physical Form: Crystalline Density: 7.9 Melting Point: 1158° Boiling Point: 2200° Solubility: Insoluble in water UN No.: 3288 MDL No.: MFCD00049611

Kaava: F₃Tm
Kiehumispiste: 2200 °C
Sulamispiste: 1158 °C
Tiheys: 7,9
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00049611
CAS-numero: 13760-79-7
EINECS: 237-353-7
UN: 3288
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR