Herasafe™ 2030i Biological Safety Cabinets
Toimittaja: Thermo Scientific
51032324 51032325 51032326 51032327 51032328 51032329 51032330 51032331
135-2667EA 19110 EUR
135-2667 135-2666 135-2663 135-2662 135-2665 135-2664 135-2661 135-2660
Herasafe™ 2030i Biological Safety Cabinets
Vetokaapit ja kotelot Biologiset suojakaapit
HeraSafe 2030i biological safety cabinets have a large interior workspace made of 316 non-corrosive stainless steel for incorporation of instruments and process optimisation devices, including robotic arms.
  • Controlled passcode access
  • Interior LED lights
  • Event and alert messaging logs
  • Standard smart ports
  • Padded armrests
  • Self-adjusting germicidal UV light intensity
Equipped with smart self-monitoring safety features, intuitive graphic touch screen interface and built-in connectivity to preserve important performance data. Electronic sash with fullyMore Product Information