Horizontal laminar flow cabinets, Aeolus Series, Telstar
Toimittaja: Azbil Telstar Technologies
1511967 1511968 1511969 1511970
135-2608EA 7060 EUR
135-2608 135-2609 135-2610 135-2611
Horizontal laminar flow cabinets, Aeolus Series, Telstar
Vetokaapit ja kotelot Laminaarivirtauskaapit
Aeolus H Serie is a high quality horizontal laminar flow workbench range for handling non-hazardous materials that require a particle free environment. These cabinets offer the highest product protection. Aeolus H horizontal laminar flow benches, have been specially designed to work in hospitals, pharmacies, IVF centres, food control, horticulture in vitro cultures, and also in electronics, optics, micromechanics, plastic Industries, etc. Horizontal laminar flow systems allow operation in sterile and particle free conditions thanks to the continuous flushing of the working area by means of aMore Product Information