Vertical laminar flow cabinets, Aeolus Series
Toimittaja: Azbil Telstar Technologies
1511963 1511964 1511965 1511966
135-2604EA 8450 EUR
135-2604 135-2605 135-2606 135-2607
Vertical laminar flow cabinets, Aeolus Series
Vetokaapit ja kotelot Laminaarivirtauskaapit
Aeolus V Serie is a high quality vertical laminar flow workbench range for handling non-hazardous materials that require a particle free environment. These cabinets offer the highest product protection. Aeolus V vertical laminar flow benches, have been specially designed to work in hospitals, pharmacies, IVF centers, food control, horticulture in vitro cultures and also in electronics, optics, micromechanics, plastic industries, etc
  • Energy saving
  • Compact external size without parts protruding from the main body; easy to clean
  • LED lighting provides uniform