Cabinets for chemicals, CS-CLASSIC
Toimittaja: asecos
30494040 30494041 30494046 30494047 30497040 30497041 30497046 30497047 30497050
135-1880EA 1250 EUR
135-1880 MSGA30494041 135-1881 MSGA30494047 MSGA30497040 MSGA30497041 MSGA30497046 MSGA30497047 MSGA30497050
Cabinets for chemicals, CS-CLASSIC
Kalusteet Säilytyskaapit
For safe and approved storage of non-flammable chemicals and toxic substances in workrooms.
  • Door variants: wing doors, wing doors with glass openings, space saving folding doors
  • Natural ventilation at the bottom (vent openings at the cabinet bottom)
  • Connection (NW 75) to a technical exhaust system on the top
  • Double-walled door construction
  • High-quality, scratch-resistant structured paint finish
  • Safety cylinder lock integrated in the turning handle
  • Adjusting aids to compensate for uneven floor
Outer body made of sheet steel steel with highgradeMore Product Information