Reniumtrikloridi

Toimittaja: Thermo Fisher Scientific
Varoitus

13230.01 13230.03
13230.01EA 385 EUR
13230.01 13230.03
Reniumtrikloridi
Reniumtrikloridi

CAS No.: 13569-63-6 Molecular Formula: ReCl3 Formula Weight: 292.56 Physical Form: Powder Sensitivity: Moisture Sensitive Odour: Odourless MDL No.: MFCD00011196

Kaava: Cl₃Re
MW: 292.57 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011196
CAS-numero: 13569-63-6
EINECS: 236-987-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR