Lutetium(III) 2,4-pentanedionate ≥99.9% (REO, rare earth oxide basis), REacton®

Toimittaja: Thermo Fisher Scientific

13189.03 13189.06
13189.03EA 411 EUR
13189.03 13189.06
Lutetium(III) 2,4-pentanedionate ≥99.9% (REO, rare earth oxide basis), REacton®
Lutetium(III) 2,4-pentanedionate

Purity: 99.9% CAS No.: 17966-84-6 Molecular Formula: C15H21LuO6 Formula Weight: 472.30 Physical Form: Powder Sensitivity: Hygroscopic MDL No.: MFCD00078030

Kaava: C₁₅H₂₁LuO₆
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00078030
CAS-numero: 17966-84-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR