Manganese(IV) telluride ≥99.9% (metals basis)

Toimittaja: Thermo Fisher Scientific
Varoitus

13134.09 13134.18
13134.09EA 247 EUR
13134.09 13134.18
Manganese(IV) telluride ≥99.9% (metals basis)
Manganese(IV) telluride

Purity: 99.9% CAS No.: 12032-89-2 Molecular Formula: MnTe2 Formula Weight: 310.14 Physical Form: 6mm & Down Pieces Odour: Odourless MDL No.: MFCD00064703

Kaava: MnTe₂
MW: 310.14 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00064703
CAS-numero: 12032-89-2
EINECS: 234-783-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR