Chemistry pump units, VARIO® select
Toimittaja: VACUUBRAND
20700200 20733550 20737550 20738450
131-0012EA 4464 EUR
131-0012 131-0029 131-0046 131-0052
Chemistry pump units, VARIO® select
Pumput Membraanipumput
The VACUU·SELECT controller simplifies your work and allows VARIO select chemistry pumping systems to deliver unparalleled precision. The pumps run whisper quiet and respond to demand in real time by matching pumping speed to process conditions. The intuitive VACUU·SELECT controller includes pre-defined applications for all common vacuum processes.
  • Ultimate combination of speed controlled pumping and intuitive application-based control
  • Simplify lab work with the VACUU·SELECT vacuum controller with graphic user interface and pre-defined applications