Seleeni(IV)kloridi 99,5 Metals basis

Toimittaja: Thermo Fisher Scientific
Vaara

Synonyms: Selenium(IV) chloride

13096.14 13096.22
13096.14EA 318 EUR
13096.14 13096.22
Seleeni(IV)kloridi 99,5 Metals basis
Seleeni(IV)kloridi

Purity: 99.5% CAS No.: 10026-03-6 Molecular Formula: SeCl4 Formula Weight: 220.77 Physical Form: Lump Density: 2.600 Refractive Index: 1.807 Sensitivity: Moisture Sensitive Odour: Pungent Melting Point: subl. Solubility: Decomposes in water and moist air. Insoluble in liquid bromine. Sublimes when heated Merck Reference: 14,8438 UN No.: 2923 MDL No.: MFCD00011225

Kaava: Cl₄Se
MW: 220.77 g/mol
Sulamispiste: 330 (subl.)
Tiheys: 2.600
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011225
CAS-numero: 10026-03-6
EINECS: 233-053-5
UN: 2923
ADR: 8 II
Merck Index: 14,8438

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR