Pentatitanium trisilicide ≥99.5% (metals basis)

Toimittaja: Thermo Fisher Scientific
Varoitus

13087.18 13087.30
13087.18EA 311 EUR
13087.18 13087.30
Pentatitanium trisilicide ≥99.5% (metals basis)
Pentatitanium trisilicide

Purity: 99.5% CAS No.: 12067-57-1 Molecular Formula: Ti5Si3 Formula Weight: 323.76 Physical Form: -325 Mesh Powder Odour: Odourless UN No.: 3178 MDL No.: MFCD01310208

Kaava: Si₃Ti₅
Sulamispiste: 2130 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD01310208
CAS-numero: 12067-57-1
EINECS: 235-082-9
UN: 3178
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR