Peqlab peqGOLD, DNA Isolation, Bacterial DNA Mini Kit
Toimittaja: VWR Chemicals
13-3450-00EA 18.9 EUR
13-3450-00 13-3450-02
Peqlab peqGOLD, DNA Isolation, Bacterial DNA Mini Kit
Nukleiinihapporeagenssit Nucleic Acid Purification Kits and Reagents
Bacterial DNA Mini Kit for the extraction of high quality DNA from gram-positive and gram-negative bacteria. For up to 1×10⁹ cells per preparation typical yield is up to 30 µg.
  • Also suited to extractions from difficult to lyse bacteria
  • Suitable for the simultaneous processing of multiple samples
  • No need for organic extractions
  • No need for time-consuming alcohol precipitations
  • Efficient separation of inhibitors
Toimitustiedot: Kit contains peqGOLD DNA columns, 2 ml collection tubes, Proteinase K, TL buffer, BL buffer, HBC buffer, glass beads S, RNAse A, Lysozyme, DNA wash and elution buffer.