Rauta ≥99.99% (metals basis), paloja low oxygen, vacuum remelted

Toimittaja: Thermo Fisher Scientific

12958.14 12958.22
12958.14EA 598 EUR
12958.14 12958.22
Rauta ≥99.99% (metals basis), paloja low oxygen, vacuum remelted
Rauta

Purity: 99.99% CAS No.: 7439-89-6 Physical Form: Oxygen and Nitrogen analysis included MDL No.: MFCD00010999

Kaava: Fe
Kiehumispiste: 3000 °C (1013 hPa)
Sulamispiste: 1535 °C
Tiheys: 7,86 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00010999
CAS-numero: 7439-89-6
EINECS: 231-096-4
Merck Index: 13,05109

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR