Nikkeli ≥99.97% (metals basis), paloja low oxygen, vacuum remelted

Toimittaja: Thermo Fisher Scientific
Vaara

12957.14 12957.22
12957.14EA 627 EUR
12957.14 12957.22
Nikkeli ≥99.97% (metals basis), paloja low oxygen, vacuum remelted
Nikkeli

Purity: 99.97% CAS No.: 7440-02-0 Physical Form: Oxygen and Nitrogen analysis included Odour: Odourless MDL No.: MFCD00011137

Kaava: Ni
Kiehumispiste: 3007 °C (1013 hPa)
Sulamispiste: 1453 °C
Tiheys: 8,9 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011137
CAS-numero: 7440-02-0
EINECS: 231-111-4
UN: 3089
ADR: 4.1,III
Merck Index: 12,06582

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR