Molybdeenitrioksidi ≥99.9995% (metals basis excluding W), Puratronic®

Toimittaja: Thermo Fisher Scientific
Vaara

Synonyms: Molybdenum trioxide

12930.22 12930.14 12930.36
12930.22EA 1330 EUR
12930.22 12930.14 12930.36
Molybdeenitrioksidi ≥99.9995% (metals basis excluding W), Puratronic®
Molybdeenitrioksidi

Purity: 99.9995% CAS No.: 1313-27-5 Molecular Formula: MoO3 Formula Weight: 143.94 Physical Form: Powder Density: 4.692 Appearance: White to grey Odour: Odourless Melting Point: 795° Boiling Point: 1155° subl. Merck Reference: 14,6239 UN No.: 3288 MDL No.: MFCD00003469

Kaava: MoO₃
MW: 143.94 g/mol
Kiehumispiste: 1155 °C (subl.)
Sulamispiste: 795
Tiheys: 4.692
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00003469
CAS-numero: 1313-27-5
EINECS: 215-204-7
UN: 3288
ADR: 6.1 III
Merck Index: 14,6239

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR