Autoclavable Bags
Toimittaja: Greiner Bio-One
643401_500 644201_500 646401_300 649401_200 644401_500
129-9721EA 387 EUR
129-9721 GREI644201_500 129-9723 129-9724 129-9722
Autoclavable Bags
Pussit ja sulkijat Jätepussit, -säkit
These bags are designed specifically for the disposal of contaminated laboratory waste, that first requires steam sterilisation.
  • Bags for autoclaving and oven - resistant up to 160 ° C in an oven and 134 ° C in an autoclave
  • Suitable for hot air sterilisers