Autoclavable Stopper Bowl Covers, DuPont™ Tyvek®
Toimittaja: KEYSTONE CLEANROOM PRODUCTS
BWLT2A20 BWLT2A24 BWLT2A26 BWLT2A30 BWLT2A34 BWLT2A36 BWLT2A38 BWLT2A42 BWLT2A46 BWLT2A48 BWLT2A54 BWLT2A56 BWLT2A60 BWLT2A64 BWLT2A72 BWLT2A90 BWLT2A120 BWLT2A144 BWLT2A52 BWLT2A32 BWLT2A36-2PLY8
129-2880EA 4610 EUR
129-2880 129-2881 129-2882 129-2883 129-2885 129-2886 129-2887 129-2888 129-2889 129-2890 129-2892 129-2893 129-2894 129-2895 129-2896 129-2897 129-2878 129-2879 129-2891 129-2884 129-3989
Autoclavable Stopper Bowl Covers, DuPont™ Tyvek®
Kuoret
Save time and standardise activities in equipment preparation for sterilisation. Leave in place during stopper bowl installation to prevent microbial and particulate contamination, and while remaining fill line set-up activities are performed.
  • Manufactured and packed in a controlled cleanroom environment
  • Reduces labour costs
  • Individually poly bagged
  • Simplifies conformance when wrapping to SOP's
  • Steam sterilisation indicator - autoclavable at 121 to 125 °C
  • Elastic to fit a range of diameters
  • Tear and puncture resistant, hydrophobic, and lint-free Tyvek® 1422