Tyvek® 1073B autoclavable wrappers
Toimittaja: KEYSTONE CLEANROOM PRODUCTS
MGTVKSHEET0203 MGTVKSHEET0303 MGTVKSHEET0306 MGTVKSHEET0404 MGTVKSHEET0406 MGTVKSHEET0518 MGTVKSHEET0536 MGTVKSHEET06 MGTVKSHEET0606 MGTVKSHEET0707 MGTVKSHT075075 MGTVKSHEET0808 MGTVKSHEET0810 MGTVKSHEET0836 MGTVKSHEET0909 MGTVKSHEET0911 MGTVKSHEET10 MGTVKSHEET1010 MGTVKSHEET1036 MGTVKSHEET1212 MGTVKSHEET14 MGTVKSHEET1515 MGTVKSHEET1616 MGTVKSHEET1818 MGTVKSHEET1836 MGTVKSHEET1860 MGTVKSHEET2020 MGTVKSHEET2424 MGTVKSHEET2430 MGTVKSHEET3030 MGTVKSHEET2854 MGTVKSHEET3636 MGTVKSHEET4040 MGTVKSHEET4080 MGTVKSHEET5454 MGTVKSHEET6060 MGTVKSHT60140
129-3137EA 726 EUR
129-3137 129-3138 129-3139 129-3140 129-3141 129-3142 129-3143 129-3144 129-3145 129-3146 129-3103 129-3147 129-3148 129-3149 129-3150 129-3151 129-3152 129-3122 129-3084 129-3085 129-3086 129-3087 129-3088 129-3089 129-3090 129-3091 129-3092 129-3093 129-3094 129-3096 129-3095 129-3097 129-3098 129-3099 129-3101 129-3102 129-3104
Tyvek® 1073B autoclavable wrappers
Pullon suojat, alumiinia
Medical grade 1073B Tyvek® wrappers improve the microbial barrier and cleanliness of your sterilisation wrap.
  • Manufactured and packaged in a controlled cleanroom environment
  • Autoclavable at 121 to 125 °C
  • Tear and puncture resistant, hydrophobic and lint-free Tyvek® 1073B
Medical grade Tyvek® 1073B wrappers are an improvement on cellulose or SMS based wrapping materials for steam sterilisation. Eliminate the wasted time and expense of cutting larger square sheets of sterilisation wrap to the desired sizes. Round shaped wrappers are an ideal way toMore Product Information