Autoclave Bags with Steam Indicator, Heat Seal, Tyvek® 1073B
Toimittaja: KEYSTONE CLEANROOM PRODUCTS
COVMGTVKUB1X14 COVMGTVKUB3X6 COVMGTVKUB3X36 COVMGTVKUB5X25 COVMGTVKUB6X8 COVMGTVKUB6X16 COVMGTVKUB6X22 COVMGTVKUB6X34 COVMGTVKUB8X9 COVMGTVKUB8X12 COVMGTVKUB8X14 COVMGTVKUB8X30 COVMGTVK10X12 COVMGTVK10X20 COVMGTVK10X25 COVMGTVK12X12 COVMGTVK12X16 COVMGTVK12X18 COVMGTVK12X24 COVMGTVK12X32 COVMGTVK12X34 COVMGTVK14X22 COVMGTVK14X28 COVMGTVK14X42 COVMGTVK15X18 COVMGTVK16X24 COVMGTVK16X32 COVMGTVK16X36 COVMGTVK16X44 COVMGTVK18X22 COVMGTVK18X24 COVMGTVK18X26 COVMGTVK18X30 COVMGTVK18X40 COVMGTVK18X68 COVMGTVK20X20 COVMGTVK20X32 COVMGTVK24X24 COVMGTVK24X44 COVMGTVK25X37 COVMGTVK26X28 COVMGTVK26X30 COVMGTVK30X30 COVMGTVK30X22 COVMGTVK30X32 COVMGTVK32X34 COVMGTVK34X34 COVMGTVK34X40 COVMGTVK36X36 COVMGTVK36X42 COVMGTVK36X48 COVMGTVK36X60 COVMGTVK38X40 COVMGTVK38X48 COVMGTVK40X48 COVMGTVK42X46 COVMGTVK48X28 COVMGTVK48X48 COVMGTVK48X64
129-2738EA 545 EUR
129-2738 129-2740 129-2739 129-2742 129-2746 129-2743 129-2744 129-2745 129-2750 129-2747 129-2748 129-2749 129-2697 129-2698 129-2699 129-2700 129-2701 129-2702 129-2703 129-2704 129-2705 129-2706 129-2707 129-2708 129-2709 129-2711 129-2712 129-2713 129-2714 129-2715 129-2716 129-2717 129-2682 129-2663 129-2662 129-2660 129-2659 129-2658 129-2657 129-2656 129-2655 129-2650 129-2647 129-2649 129-2648 129-2651 129-2652 129-2653 129-2654 129-2664 129-2665 129-2666 129-2667 129-2668 129-2671 129-2673 129-2674 129-2675 129-2676
Autoclave Bags with Steam Indicator, Heat Seal, Tyvek® 1073B
Pussit ja sulkijat Sterilointipussit
Tyvek® 1073B bags are ideal for steam sterilisation of pharmaceutical components such as vial and syringe stoppers and caps as well as heavy equipment parts with sharp edges.
  • Manufactured and packed in a controlled cleanroom environment
  • Steam sterilisation indicator - autoclavable at 121 to 125 °C
  • Tear and puncture resistant, hydrophobic and lint-free
  • 100% medical grade Tyvek® 1073B allows fast steam penetration
  • Excellent impact resistance with exceptional microbial barrier
  • Consistently strong factory seals
100% medical and pharmaceutical grade Tyvek®More Product Information