Rigid Conductive Boxes
Toimittaja: DESCO
241030 241050 241085
129-2619EA 4.1 EUR
129-2619 129-2620 129-2621
Rigid Conductive Boxes
Laatikot ESD-laatikot
Desco Europe Rigid Conductive Boxes are marked with the ESD protective symbol. Boxes can be supplied with or without high density foam for insertion of component leads or low density foam which acts as a cushioning material.
  • Provide good ESD and mechanical protection
  • Made of conductive polypropylene; Rs: 1×10² to <1×10⁵ Ω per IEC 61340-2-3
  • Strong box moulded in one-piece
  • Supplied with/without foam lining
  • Top foam: 6 mm thick low density foam (FX)
  • Bottom foam: 6 mm thick high density foam (HD)
Sertifikaatit: Meets EN 61340-5-1 and Packaging standard IEC 61340-5-3.