Autoclavable muff covers, with drawstring, Tyvek®
Toimittaja: KEYSTONE CLEANROOM PRODUCTS
MFT2A05D MFT2A07D MFT2A09D MFT2A12D MFT2A16D
129-1769EA 6050 EUR
129-1769 129-1770 129-1771 129-1772 129-1773
Autoclavable muff covers, with drawstring, Tyvek®
Kuoret
Save time and standardise activities in equipment preparation for sterilisation.
  • Manufactured and packed in a controlled cleanroom environment
  • Reduces labour costs
  • Simplifies the conformance to wrapping SOP's
  • Conveniently packed: 100 per sealed poly bag
  • Autoclavable at 121 to 125 °C
  • Tear and puncture resistant
  • Hydrophobic
  • Lint-free
Tyvek® muffs provide a convenient means to cover flasks, beakers, containers, media bottles, valves, pipes and hoses. These closures are breathable and can be used to prevent particulates and microbial contamination fromMore Product Information