VWR®, High Performance Blender Bags
Toimittaja: VWR Collection
129-1102EA 95 EUR
129-1102 129-1103 129-1104 129-1105 129-1106
VWR®, High Performance Blender Bags
Pussit ja sulkijat Blender-pussit
Complete range of bags, sterile and alimentary certified, for safe and quick preparation of solid samples before microbiological and chemical analysis. Bags are resistant for three minutes blending (frequency 4 t/second). They fit all conventional 400 ml laboratory blenders, and are adapted to all samples.
  • Can resist to high temperature from –40 to +80 °C
  • Can be frozen
  • Gamma irradiated: No risk of cross-contamination (the red sticker on every box serves as a qualitative indicator of gamma ray processing)
  • Colour-change label when sterile: Red colour, for