Hand held scrapers
Toimittaja: MICRONOVA
HHSWH-8 HHSWH-10
129-0608EA 39.4 EUR
129-0608 129-0609
Hand held scrapers
Kaapimet
This hand held scraper tool is made with a seamless, one piece injection-mould that prevents bacterial growth. Designed for residue and build-up removal in clean and controlled environments.
  • Injection-moulded, seamless construction
  • Will not scratch metal surfaces
  • Made from FDA approved polypropylene
  • Suitable for indirect food contact
  • Handle shaped for strong grip
  • Autoclavable
The scraper is chemical-resistant and will not rust or corrode.