Flathead mop covers and hardware, Slimline™
Toimittaja: MICRONOVA
SLSC6-9 SLSC6-14 SLSC6-18 SLSC6-14IR SLSC6AA-14 SLSC6-9IR SLSC6-18IR SMC6-30B
129-0550EA 35.8 EUR
129-0550 129-0548 129-0549 129-0247 129-0636 129-0630 129-0629 MICRSMC6-30B
Flathead mop covers and hardware, Slimline™
Siivousvälineet
A series of elasticised slipcovers, with urethane foam interiors, that are ideal for fine cleaning and sanitising all surfaces and equipment in the cleanroom. Available without a foam interior or gamma irradiated for aseptic environments.
  • Covers are easy to change and fit securely over mop frame
  • Fully encased Urethane Foam layer for ample disinfectant dispersal
  • Single-ply covers available without foam interior
  • Fits on QPSL plastic frame or QDSL stainless steel frame
  • Suitable for use with strong disinfectants and repeated autoclaving
The MicronovaMore Product Information