Mops, PowerHead™
Toimittaja: MICRONOVA
PHU-124 PHU-154 PH6-154 PH6-150
129-0523EA 38.8 EUR
129-0523 129-0525 129-0521 MICRPH6-150
Mops, PowerHead™
Siivousvälineet
Mop head is constructed of polyester-covered foam mop heads are available both nonsterile and sterile for critical cleaning. Absorbent foam mop head is for pharmaceutical production and buffer areas. Wide-pore foam mop head and dense urethane foam mop head are for cleanroom support and maintenance areas.
  • Stainless steel PowerHead™ handles hold and wring the various mop heads
  • Autoclavable
  • A quick-disconnect fitting on the handle allows the user to customize length with single-piece and extendable universal handles
Tilaustiedot: All components are ordered separately.