Bags, radiation bin box, B-Safe
Toimittaja: Alpha Packaging
526081 526083 526084 526085
129-0407EA 273 EUR
129-0407 129-0409 129-0410 129-0411
Bags, radiation bin box, B-Safe
Pussit ja sulkijat Jätepussit, -säkit
The bags are printed with yellow radioactive warning symbol and a maximum fill line. PE,125 µm (500 gauge), clear.
  • Heat sealed for leak protection
  • Each bag is box-shaped and has a double-string neck-pull