Self-Seal Biohazard Bags
Toimittaja: Alpha Packaging
121103 121104 121105
129-1931EA 193 EUR
129-1931 129-0398 129-0399
Self-Seal Biohazard Bags
Pussit ja sulkijat Jätepussit, -säkit
Tough, yellow biohazard bags, non autoclavable. Supplied in benchtop dispenser boxes. PE, 62,5 µm (250 gauge).
  • Peel-off permanent adhesive seal
  • Remove backing and seal for safer transportation/disposal
  • Biohazard warning in English, German and French, black printing on yellow background