Self-wringing cleanroom roller mops, Roll-O-Matic®
Toimittaja: FHP VILEDA SCS
118771 120848 121165 119654 119657 119658 118550 118549 122910
129-0195EA 229 EUR
129-0195 129-0270 129-0271 VILE119654 VILE119657 VILE119658 141-1440 141-1439 141-1441
Self-wringing cleanroom roller mops, Roll-O-Matic®
Siivousvälineet
Roll-O-Matic® is the ideal tool for cleaning or disinfecting floors, walls and ceilings in hygiene-sensitive areas.
It has a self-wringing mechanism, and is best used with the 'pull and lift' technique recommended for cleaning and disinfecting. It’s simple to use, and elminates bulky, expensive wringers.
  • Integrated wringing mechanism for use with 'pull and lift' technique
  • Coloured clips on the handles allow for colour coding of equipment
  • Stainless steel handles are compatible with gamma and autoclave sterilisation
  • Straight head design allows the user to